Perfluoro-1-octene

Supplier: APOLLO SCIENTIFIC
Warning

PC3331-1G PC3331-5G
77428-760EA 110.04 USD
77428-760 77428-762
Perfluoro-1-octene
Perfluoro-1-octene
Formula: C₈F₁₆
Boiling Pt: 102…105 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00236708
CAS Number: 559-14-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR