Methyl-4-hydroxybenzoate 98%

Supplier: APOLLO SCIENTIFIC

OR922886-100G OR922886-1KG OR922886-500G
77425-076EA 21.26 USD
77425-076 77425-078 77425-080
Methyl-4-hydroxybenzoate 98%
Methyl 4-hydroxybenzoate
Formula: HOC₆H₄CO₂CH₃
Melting Pt: 125…128 °C
Storage Temperature: Ambient
MDL Number: MFCD00002352
CAS Number: 99-76-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR