3-Methyl-3-pentanol 98%

Supplier: APOLLO SCIENTIFIC

Synonyms: Diethylmethylcarbinol

OR922588-100G OR922588-25G OR922588-5G
77411-806EA 241.46 USD
77411-806 77411-808 77411-810
3-Methyl-3-pentanol 98%
3-Methyl-3-pentanol
Formula: C₆H₁₄O
MW: 102.18 g/mol
Boiling Pt: 123
Melting Pt: -53
Storage Temperature: Ambient
MDL Number: MFCD00004483
CAS Number: 77-74-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR