5-Fluoro-o-toluidine 97%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
PC49276-100G
PC49276-1KG
PC49276-500G
77415-900EA
37.28
USD
77415-900
77415-902
77415-904
5-Fluoro-o-toluidine 97%
5-Fluoro-o-toluidine
Formula:
C₇H₈FN Boiling Pt: 98…100 °C/15 mmHg Melting Pt: 37…40 °C Flash Pt: 90 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007764 CAS Number: 367-29-3 EINECS: 206-689-6 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...