3,5-Difluorobenzylamine 97%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
PC2738D-10G
PC2738D-1G
PC2738D-5G
77413-806EA
41.78
USD
77413-806
77413-808
77413-810
3,5-Difluorobenzylamine 97%
3,5-Difluorobenzylamine
Formula:
C₇H₇F₂N Boiling Pt: 184 °C Density: 1.21 g/cm³ (25 °C) Flash Pt: 73 °C Storage Temperature: Ambient |
MDL Number:
MFCD00061244 CAS Number: 90390-27-5 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...