2,5-Difluorobenzylamine 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: (2,5-Difluorophenyl)methanamine

PC2736-100G PC2736-25G PC2736-5G
77413-794EA 221.08 USD
77413-794 77413-796 77413-798
2,5-Difluorobenzylamine 95%
2,5-Difluorobenzylamine
Formula: C₇H₇F₂N
Boiling Pt: 76…78 °C/15 mmHg
Density: 1.222 g/cm³ (25 °C)
Flash Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD00010143
CAS Number: 85118-06-5
EINECS: 285-658-9
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR