2,4-Dibromotoluene 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2,4-Dibromo-1-methylbenzene

OR2172-25G OR2172-5G
77406-714EA 37.7 USD
77406-714 77406-716
2,4-Dibromotoluene 98%
2,4-Dibromotoluene
Formula: C₇H₆Br₂
MW: 249.93 g/mol
Boiling Pt: 243 °C
Melting Pt: –10 °C
Density: 1.85 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00052985
CAS Number: 31543-75-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR