Potassium-tert-butoxide 97%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
OR11033-100G
OR11033-1KG
OR11033-500G
77403-258EA
96.78
USD
77403-258
77403-260
77403-262
Potassium-tert-butoxide 97%
Potassium-tert-butoxide
Formula:
C₄H₉KO Melting Pt: 255 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00012162 CAS Number: 865-47-4 EINECS: 212-740-3 UN: 3206 ADR: 4.2+8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...