3-Methyladenine

Supplier: APOLLO SCIENTIFIC
Warning

OR470734-10MG OR470734-1G OR470734-5G
77421-918EA 21.4 USD
77421-918 77421-920 77421-922
3-Methyladenine
3-Methyladenine
Formula: C₆H₇N₅
Storage Temperature: Ambient
MDL Number: MFCD00010531
CAS Number: 5142-23-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR