Piperazine 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR13160-100G OR13160-500G
77404-562EA 20.26 USD
77404-562 77404-564
Piperazine 97%
Piperazine
Formula: C₄H₁₀N₂
Boiling Pt: 145…148 °C
Melting Pt: 108…113 °C
Density: 1.1 g/cm³ (25 °C)
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00005953
CAS Number: 110-85-0
EINECS: 203-808-3
UN: 2579
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR