Deuterium oxide (>99.9% D)

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Heavy waterWater-D2

DE50K-100G
77401-254EA 202.91 USD
77401-254
Deuterium oxide (>99.9% D)
Deuterium oxide
Formula: D₂O
MW: 20 g/mol
Boiling Pt: 101.4 °C
Melting Pt: 3.8 °C
Density: 1.12 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00044636
CAS Number: 7789-20-0
EINECS: 232-148-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR