4-Aminoindole

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Indol-4-ylamine, 4-Indolamine, 4-Amino-1H-indole

OR7782-1G OR7782-5G
77424-328EA 75.4 USD
77424-328 77424-330
4-Aminoindole
4-Aminoindole
Formula: C₈H₈N₂
MW: 132.16 g/mol
Melting Pt: 108
Storage Temperature: Refrigerator
MDL Number: MFCD01076559
CAS Number: 5192-23-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR