3-Hydroxypropiononitrile

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Ethylene Cyanohydrin, 2-Cyanoethanol, 3-Hydroxypropionitrile, Hydracrylonitrile, β-Hydroxypropiononitrile

OR3049-1KG OR3049-250G
77416-976EA 190.52 USD
77416-976 77416-978
3-Hydroxypropiononitrile
3-Hydroxypropiononitrile
Formula: C₃H₅NO
Boiling Pt: 227…228 °C
Melting Pt: –46 °C
Density: 1.041 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00002826
CAS Number: 109-78-4
EINECS: 203-704-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR