Deuterium chloride 20% in D₂O (99.95% D)
Supplier: APOLLO SCIENTIFIC
|
Danger
|
DE760B-25ML
77401-296EA
210.12
USD
77401-296
Deuterium chloride 20% in D₂O (99.95% D)
Deuterium chloride
Formula:
DCl MW: 37.45 g/mol Boiling Pt: 108 °C Density: 1.2 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00044642 CAS Number: 7698-05-7 EINECS: 231-715-8 UN: 1789 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...