Deuterium chloride 20% in D₂O (>99.0% D)

Supplier: APOLLO SCIENTIFIC
Danger

DE47-10G
77401-248EA 52.53 USD
77401-248
Deuterium chloride 20% in D₂O (>99.0% D)
Deuterium chloride
Formula: DCl
MW: 37.45 g/mol
Boiling Pt: 108 °C
Density: 1.11 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00044642
CAS Number: 7698-05-7
EINECS: 231-715-8
UN: 1789
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR