Cyclopropanemethylamine 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR1937-25G OR1937-5G
77406-444EA 75.4 USD
77406-444 77406-446
Cyclopropanemethylamine 97%
Cyclopropanemethylamine
Formula: C₄H₉N
Boiling Pt: 84…85 °C
Density: 0.82 g/cm³ (25 °C)
Flash Pt: 2 °C
Storage Temperature: Ambient
MDL Number: MFCD00037147
CAS Number: 2516-47-4
EINECS: 219-737-6
UN: 2733
ADR: 3+8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR