Bromocyclobutane 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Cyclobutyl bromide

OR41168-25G OR41168-5G
77409-610EA 41.78 USD
77409-610 77409-612
Bromocyclobutane 98%
Bromocyclobutane
Formula: C₄H₇Br
MW: 135 g/mol
Boiling Pt: 107…108 °C
Density: 1.434 g/cm³ (25 °C)
Flash Pt: 22 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001317
CAS Number: 4399-47-7
EINECS: 224-530-9
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR