Quinoline 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR45105-100G OR45105-25G OR45105-500G
77409-760EA 66.22 USD
77409-760 77409-762 77409-764
Quinoline 98%
Quinoline
Formula: C₉H₇N
Boiling Pt: 236…238 °C
Melting Pt: –17…–13 °C
Density: 1.093 g/cm³ (25 °C)
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00006736
CAS Number: 91-22-5
EINECS: 202-051-6
UN: 2656
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR