4-Bromo-o-xylene

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Bromo-1,2-dimethylbenzene

OR1071-100G OR1071-25G
77402-902EA 83.54 USD
77402-902 77402-904
4-Bromo-o-xylene
4-Bromo-o-xylene
Formula: C₈H₉Br
Boiling Pt: 214…215 °C
Density: 1.34 g/cm³ (25 °C)
Flash Pt: 80 °C
Storage Temperature: Ambient
MDL Number: MFCD00000071
CAS Number: 583-71-1
EINECS: 209-518-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR