1,3-Dichloro-4-fluorobenzene 99%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2,4-Dichlorofluorobenzene, 2,4-Dichloro-1-fluorobenzene

PC2398M-100G PC2398M-25G
77413-622EA 68.26 USD
77413-622 77413-624
1,3-Dichloro-4-fluorobenzene 99%
1,3-Dichloro-4-fluorobenzene
Formula: C₆H₃Cl₂F
Boiling Pt: 172…174 °C
Melting Pt: –23 °C
Density: 1.409 g/cm³ (25 °C)
Flash Pt: 37 °C
Storage Temperature: Ambient
MDL Number: MFCD00000580
CAS Number: 1435-48-9
EINECS: 406-160-1
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR