4-Chlorobenzaldehyde 98%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR12011-100G
OR12011-1KG
OR12011-250G
77404-114EA
32.6
USD
77404-114
77404-116
77404-118
4-Chlorobenzaldehyde 98%
4-Chlorobenzaldehyde
Formula:
C₇H₅ClO Boiling Pt: 213…214 °C Melting Pt: 45…50 °C Density: 1.196 g/cm³ (25 °C) Flash Pt: 87 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003379 CAS Number: 104-88-1 EINECS: 203-247-4 UN: 3077 ADR: 9+EHS,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...