4-Bromo-2-fluorophenol 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-Bromo-3-fluoro-4-hydroxybenzene

PC1429E-25G PC1429E-5G
77427-100EA 23.44 USD
77427-100 77427-102
4-Bromo-2-fluorophenol 98%
4-Bromo-2-fluorophenol
Formula: C₆H₄BrFO
MW: 191 g/mol
Boiling Pt: 90
Density: 1.744 g/cm³ (25 °C)
Flash Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD00011722
CAS Number: 2105-94-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR