1,6-Diiodoperfluorohexane 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Perfluoro-1,6-diiodohexane

PC2967-25G PC2967-5G
77428-040EA 35.02 USD
77428-040 77428-042 77428-044
1,6-Diiodoperfluorohexane 98%
1,6-Diiodoperfluorohexane
Formula: C₆F₁₂I₂
Boiling Pt: 114…116 °C/100 mmHg
Melting Pt: 25 °C
Density: 2.357 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00042264
CAS Number: 375-80-4
EINECS: 206-794-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR