Trifluoroacetic acid 99,9%, Biochemical Grade
Supplier: APOLLO SCIENTIFIC
|
Danger
|
PC7161-1KG
PC7161-250G
77417-668EA
74.38
USD
77417-668
77417-670
Trifluoroacetic acid 99,9%, Biochemical Grade
Trifluoroacetic acid
Suitable for protein sequence analysis.
Formula:
C₂HF₃O₂ Boiling Pt: 72.4 °C Melting Pt: –15.4 °C Density: 1.489 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004169 CAS Number: 76-05-1 EINECS: 200-929-3 UN: 2699 ADR: 8,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...