2,2,2-Trichloroethanol

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2,2,2-Trichloroethan-1-ol

OR40732-100G OR40732-500G
77421-208EA 89.66 USD
77421-208 77421-210
2,2,2-Trichloroethanol
2,2,2-Trichloroethanol
Formula: C₂H₃Cl₃O
Boiling Pt: 152…154 °C
Melting Pt: 17.8 °C
Density: 1.56 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004677
CAS Number: 115-20-8
EINECS: 204-071-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR