Benzene-[D₆] (>99.50% D)

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Benzene-[²H₆]

DE30L-100ML
77401-216EA 413.03 USD
77401-216
Benzene-[D₆] (>99.50% D)
Benzene-[D6]

(H2O)<0.02%

Formula: C₆D₆
MW: 84.07 g/mol
Boiling Pt: 79.1 °C
Melting Pt: 6.8 °C
Density: 0.95 g/cm³ (25 °C)
Flash Pt: –11 °C
Storage Temperature: Ambient
MDL Number: MFCD00003010
CAS Number: 1076-43-3
UN: 1114
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR