Acetone-[D₆] (>99,8% D)

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Propanone-D6DMK-D6HexadeuteroacetoneAcetone-d6Dimethyl ketone-D6Acetone-[²H₆]

DE10H-10G
77401-174EA 40.17 USD
77401-174
Acetone-[D₆] (>99,8% D)
Acetone-[D6]

(HDO+D2O)<0.02%

Formula: D₃CC(O)CD₃
MW: 64.03 g/mol
Boiling Pt: 55.5 °C
Melting Pt: –93.8 °C
Density: 0.872 g/cm³ (25 °C)
Flash Pt: –19 °C
Storage Temperature: Ambient
MDL Number: MFCD00044635
CAS Number: 666-52-4
EINECS: 211-563-9
UN: 1090
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR