(S)-(-)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid 99%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (S)-(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetic acid, (2S)-(-)-2-Methoxy-2-phenyl-3,3,3-trifluoropropanoic acid

PC5063E-100MG PC5063E-1G PC5063E-5G
77431-002EA 34.64 USD
77431-002 77431-004 77431-006
(S)-(-)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid 99%
(S)-(-)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid
Formula: C₁₀H₉F₃O₃
MW: 234.17 g/mol
Boiling Pt: 95…97 °C/0.05 mmHg
Melting Pt: 46…49 °C
Density: 1.303 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064200
CAS Number: 17257-71-5
EINECS: 241-292-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR