3-Methoxy-5-(trifluoromethyl)benzyl alcohol 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC49338-1G PC49338-5G
77430-434EA 103.92 USD
77430-434 77430-436
3-Methoxy-5-(trifluoromethyl)benzyl alcohol 97%
3-Methoxy-5-(trifluoromethyl)benzyl alcohol
Formula: C₉H₉F₃O₂
Melting Pt: 32…35 °C
Storage Temperature: Ambient
MDL Number: MFCD09025413
CAS Number: 916420-98-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR