3,6-Bis(1,1-dimethylethyl)-10-phenyl-9-(2,4,6-trimethylphenyl)-acridinium tetrafluoroborate 95%

Supplier: APOLLO SCIENTIFIC

PC49681-100MG PC49681-1G PC49681-250MG
77430-642EA 54.4 USD
77430-642 77430-644 77430-646
3,6-Bis(1,1-dimethylethyl)-10-phenyl-9-(2,4,6-trimethylphenyl)-acridinium tetrafluoroborate 95%
3,6-Bis(1,1-dimethylethyl)-10-phenyl-9-(2,4,6-trimethylphenyl)-acridinium tetrafluoroborate
Formula: C₃₆H₄₀N·BF₄
Melting Pt: 277.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD31381908
CAS Number: 1810004-87-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR