2-(Difluoromethyl)benzoic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC48345-1G PC48345-250MG PC48345-5G
77430-216EA 41.78 USD
77430-216 77430-218 77430-220
2-(Difluoromethyl)benzoic acid 97%
2-(Difluoromethyl)benzoic acid
Formula: C₈H₆F₂O₂
Melting Pt: 128…130 °C
Storage Temperature: Ambient
MDL Number: MFCD16140191
CAS Number: 799814-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR