2,6-Difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2,6-Difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-benzonitrile4-Cyano-3,5-difluorobenzeneboronic acid pinacol ester

PC408301-250MG
77429-424EA 138.02 USD
77429-424
2,6-Difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
2,6-Difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
Formula: C₁₃H₁₄BF₂NO₂
MW: 265.067 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22419275
CAS Number: 1003298-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR