2-(Trifluoroacetyl)pyrrole 99%

Supplier: APOLLO SCIENTIFIC
Warning

PC3389-5G PC3389-25G
77428-816EA 118.58 USD
77428-816 77428-812 77428-814
2-(Trifluoroacetyl)pyrrole 99%
2-(Trifluoroacetyl)pyrrole
Formula: C₆H₄F₃NO
MW: 163.100 g/mol
Melting Pt: 48…50 °C
Storage Temperature: Ambient
MDL Number: MFCD00141809
CAS Number: 2557-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR