5-Bromo-2-fluoroisonicotinic acid

Supplier: APOLLO SCIENTIFIC

PC305015-1G PC305015-250MG
77428-446EA 512.2 USD
77428-446 77428-448
5-Bromo-2-fluoroisonicotinic acid
5-Bromo-2-fluoroisonicotinic acid
Formula: C₆H₃BrFNO₂
Storage Temperature: Ambient
MDL Number: MFCD13185803
CAS Number: 1214377-40-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR