3,4-Dichloro-5-(trifluoromethyl)phenylacetic acid 97%

Supplier: APOLLO SCIENTIFIC

PC303242-1G PC303242-250MG
77428-388EA 939.34 USD
77428-388 77428-390
3,4-Dichloro-5-(trifluoromethyl)phenylacetic acid 97%
3,4-Dichloro-5-(trifluoromethyl)phenylacetic acid
Formula: C₉H₅Cl₂F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD28103374
CAS Number: 1823541-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR