2-Chloro-N-((dimethylamino)methylene)-4-fluorobenzamide

Supplier: APOLLO SCIENTIFIC
Warning

PC28280-250MG PC28280-1G
77427-892EA 25.75 USD
77427-892 77427-890
2-Chloro-N-((dimethylamino)methylene)-4-fluorobenzamide
2-Chloro-N-((dimethylamino)methylene)-4-fluorobenzamide
Formula: C₁₀H₁₀ClFN₂O
MW: 228.651 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31812107
CAS Number: 2379917-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR