(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP)

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: PyBOP1H-Benzotriazol-1-yloxytri(1-pyrrolidinyl)phosphonium hexafluorophosphate((1H-Benzo[d][1,2,3]triazol-1-yl)oxy)tri(pyrrolidin-1-yl)phosphonium hexafluorophosphate(V)

PC1208-25G PC1208-100G PC1208-500G
77426-882EA 32.96 USD
77426-882 77426-884 77426-886
(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP)
(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP)

Peptide coupling reagent.

Formula: C₁₈H₂₈F₆N₆OP₂
MW: 520.4 g/mol
Melting Pt: 154…156 °C (dec.)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00077411
CAS Number: 128625-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR