2,3-Dihydro-6-(trifluoromethyl)benzo[1,4]thiazin-3-one 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC2954E-100MG PC2954E-500MG
77428-026EA 38.26 USD
77428-026 77428-028
2,3-Dihydro-6-(trifluoromethyl)benzo[1,4]thiazin-3-one 97%
2,3-Dihydro-6-(trifluoromethyl)benzo[1,4]thiazin-3-one
Formula: C₉H₆F₃NOS
Melting Pt: 192…194 °C
Storage Temperature: Ambient
MDL Number: MFCD00178754
CAS Number: 716-82-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR