4,5-Difluoroanthranilic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Amino-4,5-difluorobenzoic acid

PC10504-100G PC10504-10G PC10504-25G
77426-678EA 222.1 USD
77426-678 77426-680 77426-682
4,5-Difluoroanthranilic acid 97%
4,5-Difluoroanthranilic acid
Formula: C₇H₅F₂NO₂
Melting Pt: 174…183 °C
Storage Temperature: Ambient
MDL Number: MFCD00077509
CAS Number: 83506-93-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR