1-Ethyl-2-nitrobenzene 99%

Supplier: APOLLO SCIENTIFIC

Synonyms: 2-Ethylnitrobenzene

OR956256-100G OR956256-25G OR956256-500G
77425-768EA 45.84 USD
77425-768 77425-770 77425-772
1-Ethyl-2-nitrobenzene 99%
1-Ethyl-2-nitrobenzene
Formula: C₈H₉NO₂
MW: 151.16 g/mol
Boiling Pt: 220
Melting Pt: -11
Storage Temperature: Ambient
MDL Number: MFCD00024294
CAS Number: 612-22-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR