4-(Trifluoromethyl)phenylboronic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-(Trifluoromethyl)benzeneboronic acid, (4-(Trifluoromethyl)phenyl)metaboric acid, Dihydroxy(4-(trifluoromethyl)phenyl)borane, (4-(Trifluoromethyl)phenyl)boronic acid

PC1111-5G PC1111-100G PC1111-25G
77426-816EA 21.26 USD
77426-816 77426-806 77426-814
4-(Trifluoromethyl)phenylboronic acid 98%
4-(Trifluoromethyl)phenylboronic acid
Formula: C₇H₆BF₃O₂
MW: 189.93 g/mol
Melting Pt: 235…238 °C
Storage Temperature: Ambient
MDL Number: MFCD00151855
CAS Number: 128796-39-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR