6-Fluoroanthranilic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Amino-6-fluorobenzoic acid

PC1071M-25G PC1071M-100G
77426-742EA 40.17 USD
77426-742 77426-744
6-Fluoroanthranilic acid 97%
6-Fluoroanthranilic acid
Formula: C₇H₆FNO₂
MW: 155.13 g/mol
Melting Pt: 165…172 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00067781
CAS Number: 434-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR