(S)-1-[3,5-Bis(trifluoromethyl)phenyl]ethanol 98%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
(S)-1-[3,5-Bis(trifluoromethyl)phenyl]ethyl alcohol, (S)-(-)-3,5-Bis(trifluoromethyl)-alpha-methylbenzyl alcohol, (1S)-(-)-1-[3,5-Bis(trifluoromethyl)phenyl]ethan-1-ol, (S)-1-(3,5-Bis(trifluoromethyl)phenyl)ethanol, (S)-3,5-Bis(trifluoromethyl)-alpha-methylbenzyl alcohol
PC0581-1G
PC0581-25G
PC0581-5G
77426-296EA
21.26
USD
77426-296
77426-298
77426-300
(S)-1-[3,5-Bis(trifluoromethyl)phenyl]ethanol 98%
(S)-1-[3,5-Bis(trifluoromethyl)phenyl]ethanol
Formula:
C₁₀H₈F₆O Boiling Pt: 83 °C/11 mmHg Storage Temperature: Ambient |
MDL Number:
MFCD03093011 CAS Number: 225920-05-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...