2-Amino-3-bromophenol 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR9739-1G OR9739-250MG OR9739-5G
77425-964EA 55.02 USD
77425-964 77425-966 77425-968
2-Amino-3-bromophenol 95%
2-Amino-3-bromophenol
Formula: C₆H₆BrNO
Melting Pt: 135…137 °C
Storage Temperature: Ambient
MDL Number: MFCD08445676
CAS Number: 116435-77-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR