2-Methoxyethyl methanesulfonate

Supplier: APOLLO SCIENTIFIC

Synonyms: Methanesulphonic Acid 2-Methoxyethyl Ester, Methanesulfonic Acid 2-Methoxyethyl Ester

OR923587-1G OR923587-25G OR923587-5G
77425-096EA 27.5 USD
77425-096 77425-098 77425-100
2-Methoxyethyl methanesulfonate
2-Methoxyethyl methanesulfonate
Formula: C₄H₁₀O₄S
Boiling Pt: 80 °C
Density: 1.25 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD02656401
CAS Number: 16427-44-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR