Methyl (R)-(-)-3-hydroxyisobutyrate 97%

Supplier: APOLLO SCIENTIFIC

OR919434-10G OR919434-1G OR919434-5G
77425-008EA 253.68 USD
77425-008 77425-010 77425-012
Methyl (R)-(-)-3-hydroxyisobutyrate 97%
Methyl (R)-(-)-3-hydroxyisobutyrate
Formula: C₅H₁₀O₃
Storage Temperature: Ambient
MDL Number: MFCD00063450
CAS Number: 72657-23-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR