3-Chlorophenyl acetylene 99%

Supplier: APOLLO SCIENTIFIC
Warning

OR6594-1G OR6594-25G OR6594-5G
77423-862EA 21.26 USD
77423-862 77423-864 77423-866
3-Chlorophenyl acetylene 99%
3-Chlorophenyl acetylene
Formula: C₈H₅Cl
Boiling Pt: 178…179 °C
Density: 1.109 g/cm³ (25 °C)
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD03839984
CAS Number: 766-83-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR