Isobutamben (isobutyl 4-aminobenzoate) 98%

Supplier: APOLLO SCIENTIFIC

Synonyms: 4-Aminobenzoic acid isobutyl ester

OR922243-1G OR922243-25G OR922243-5G
77425-050EA 21.36 USD
77425-050 77425-052 77425-054
Isobutamben (isobutyl 4-aminobenzoate) 98%
Isobutamben (Isobutyl 4-aminobenzoate)
Formula: C₁₁H₁₅NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD00017110
CAS Number: 94-14-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR