1-Naphtholbenzeine

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: alpha-Naphtholbenzein, p-Naphtholbenzein, α-Naphtholbenzein, p-Naphtholbenzeine, 4-(α-(4-Hydroxy-1-naphthyl)benzylidene)naphthalen-1(4H)-one, 1-Naphtholbenzein, Bis(4-hydroxy-1-naphthyl)phenylmethanol, 4-Hydroxy-α-(4-hydroxynaphthyl)-α-phenylnaphthalene-1-methanol, α-Naphtholbenzeine

OR70081-100G OR70081-25G
77424-012EA 498.2 USD
77424-012 77424-014
1-Naphtholbenzeine
1-Naphtholbenzeine
Formula: C₂₇H₁₈O₂
MW: 374.44 g/mol
Melting Pt: ∼ 225 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00078492
CAS Number: 145-50-6
EINECS: 205-656-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR