Diethyl ethylidenemalonate 95+%

Supplier: APOLLO SCIENTIFIC

Synonyms: Diethyl 2-ethylidenemalonate

OR59915-10G OR59915-25G
77423-332EA 73.36 USD
77423-332 77423-334
Diethyl ethylidenemalonate 95+%
Diethyl ethylidenemalonate
Formula: C₉H₁₄O₄
Boiling Pt: 115…118 °C/17 mmHg
Density: 1.019 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009145
CAS Number: 1462-12-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR